p-Toluenesulfonyl isocyanate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

T0998-25G T0998-500G T0998-100G
TCT0998-025GEA 48.44 CAD
TCT0998-025G TCT0998-500G TCT0998-100G
p-Toluenesulfonyl isocyanate ≥95.0% (by titrimetric analysis)
p-Toluenesulphonyl isocyanate

CAS Number: 4083-64-1
MDL Number: MFCD00002030
Molecular Formula: C8H7NO3S
Molecular Weight: 197.21
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 130
Melting point (°C): -2
Flash Point (°C): 145
Specific Gravity (20/20): 1.30


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: Tosyl Isocyanate

Formula: C₈H₇NO₃S
Boiling Pt: 130 °C
Melting Pt: –2 °C
Density: 1.30 (20 °C)
Flash Pt: 145 °C
MDL Number: MFCD00002030
CAS Number: 4083-64-1
EINECS: 223-810-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a73d4ebf5f846d96cd820f12c20c2f291f8094c3&vid=a73d4ebf5f846d96cd820f12c20c2f291f8094c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR