1-Iodo-2,3,5,6-tetramethylbenzene ≥98.0%
Supplier: TCI America
Synonyms:
2,3,5,6-Tetramethyliodobenzene, 3-Iodo-1,2,4,5-tetramethylbenzene
T0992-5G
T0992-1G
TCT0992-005GEA
176.96
CAD
TCT0992-005G
TCT0992-1G
1-Iodo-2,3,5,6-tetramethylbenzene ≥98.0%
1-Iodo-2,3,5,6-tetramethylbenzene
CAS Number: 2100-25-6
MDL Number: MFCD00013705
Molecular Formula: C10H13I
Molecular Weight: 260.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 290
Melting point (°C): 80
Other Product Names / Synonyms: 3-Iododurene, 2,3,5,6-Tetramethyliodobenzene
Formula:
C₁₀H₁₃I MW: 260.12 g/mol Boiling Pt: 290 °C Melting Pt: 80 °C |
MDL Number:
MFCD00013705 CAS Number: 2100-25-6 EINECS: 218-266-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...