1,2-Bis(dimethylamino)methane ≥98.0%

Supplier: TCI America

Synonyms: N,N,N',N'-Tetramethylmethylenediamine, N,N,N',N'-Tetramethyldiaminomethane

T0969-500ML T0969-100ML
TCT0969-500MLEA 470.84 CAD
TCT0969-500ML TCT0969-100ML
1,2-Bis(dimethylamino)methane ≥98.0%
1,2-Bis(dimethylamino)methane

CAS Number: 51-80-9
MDL Number: MFCD00008328
Molecular Formula: C5H14N2
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 84
Flash Point (°C): 2
Specific Gravity (20/20): 0.75

Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 85
Melting Pt: -55
Density: 0.75 (20 °C)
Flash Pt: 2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008328
CAS Number: 51-80-9
EINECS: 200-124-7
UN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1cb96ee1e3c4a6963c943e818fdd25348ebb526&vid=d1cb96ee1e3c4a6963c943e818fdd25348ebb526&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR