Ethyl tridecanoate ≥97.0%

Supplier: TCI America

T0959-5ML T0959-25ML
TCT0959-5MLEA 143.78 CAD
TCT0959-5ML TCT0959-025ML
Ethyl tridecanoate ≥97.0%
Ethyl tridecanoate

CAS Number: 28267-29-0
MDL Number: MFCD00008978
Molecular Formula: C15H30O2
Molecular Weight: 242.40
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 198
Flash Point (°C): 113
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Tridecanoic Acid Ethyl Ester

Formula: C₁₅H₃₀O₂
Boiling Pt: 198 °C
Density: 0.86 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008978
CAS Number: 28267-29-0
EINECS: 248-929-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e7fcc22a2551322d055387c85c148afb8c8f864&vid=1e7fcc22a2551322d055387c85c148afb8c8f864&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR