2,3,5-Tribromothiophene ≥98.0%
Supplier: TCI America
T0956-25G
T0956-250G
TCT0956-025GEA
143.78
CAD
TCT0956-025G
TCT0956-250G
2,3,5-Tribromothiophene ≥98.0%
2,3,5-Tribromothiophene
CAS Number: 3141-24-0
MDL Number: MFCD00014521
Molecular Formula: C4HBr3S
Molecular Weight: 320.82
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Freezing point (°C): 26
Storage Temperature: 0-10°C
Formula:
C₄HBr₃S Boiling Pt: 80 °C (0.5 mmHg) Melting Pt: 29 °C |
MDL Number:
MFCD00014521 CAS Number: 3141-24-0 EINECS: 221-544-7 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...