2,3,5-Trimethylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0942-5G T0942-25G
TCT0942-005GEA 33.18 CAD
TCT0942-005G TCT0942-25G
2,3,5-Trimethylpyrazine ≥98.0% (by GC, titration analysis)
2,3,5-Trimethylpyrazine

CAS Number: 14667-55-1
MDL Number: MFCD00006145
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 172
Flash Point (°C): 67
Specific Gravity (20/20): 0.98

Formula: C₇H₁₀N₂
Boiling Pt: 172 °C
Density: 0.98 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00006145
CAS Number: 14667-55-1
EINECS: 238-712-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fd2f50eb5b06481b3fdc3cfed0e6ea2daa58e54&vid=1fd2f50eb5b06481b3fdc3cfed0e6ea2daa58e54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR