Tetradecyltrimethylammonium bromide ≥98.0%

Supplier: TCI America
Danger

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

T0906-500G T0906-25G T0906-100G
TCT0906-500GEA 205.7 CAD
TCT0906-500G TCT0906-25G TCT0906-100G
Tetradecyltrimethylammonium bromide ≥98.0%
Tetradecyltrimethylammonium bromide

CAS Number: 1119-97-7
MDL Number: MFCD00011770
Molecular Formula: C17H38BrN
Molecular Weight: 336.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry
Other Product Names / Synonyms: Myristyltrimethylammonium Bromide, TTAB, MTAB

Formula: C₁₇H₃₈BrN
MW: 336.394 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011770
CAS Number: 1119-97-7
EINECS: 214-291-9
UN: 3261
ADR: 8 II
Merck Index: 12,06419
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82414a82dd5e03d6cf235b2461fe0afe59dd93d9&vid=82414a82dd5e03d6cf235b2461fe0afe59dd93d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR