3-Mercapto-1,2-propanediol ≥95.0%

Supplier: TCI America

Synonyms: (±)-3-Mercapto-1,2-dihydroxy propane(±)-3-Mercapto-1,2-propanediol(±)-α-Monothioglycerinealpha-Thioglycerolα-Monothioglycerol(±)-1-Thioglycerine1-ThioglycerolMonothioglycerol

T0905-25G T0905-100G T0905-500G
TCT0905-100GEA 130.54 CAD
TCT0905-25G TCT0905-100G TCT0905-500G
3-Mercapto-1,2-propanediol ≥95.0%
3-Mercapto-1,2-propanediol

CAS Number: 96-27-5
MDL Number: MFCD00004879
Molecular Formula: C3H8O2S
Molecular Weight: 108.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 139
Flash Point (°C): 113
Specific Gravity (20/20): 1.25


Other Product Names / Synonyms: 3-Mercapto-1,2-propanediol

Formula: C₃H₈O₂S
MW: 108.16 g/mol
Boiling Pt: 139 °C
Density: 1.25 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00004879
CAS Number: 96-27-5
Merck Index: 12,09471
https://ca.vwr.com/stibo/web/std.lang.all/50/03/28995003.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR