3',4',5'-Trimethoxyacetophenone ≥98.0%
Supplier: TCI America
T0898-25G
T0898-5G
TCT0898-025GEA
246.48
CAD
TCT0898-025G
TCT0898-005G
3',4',5'-Trimethoxyacetophenone ≥98.0%
3',4',5'-Trimethoxyacetophenone
CAS Number: 1136-86-3
MDL Number: MFCD00008738
Molecular Formula: C11H14O4
Molecular Weight: 210.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 224
Melting point (°C): 82
Formula:
C₁₁H₁₄O₄ Boiling Pt: 224 °C Melting Pt: 82 °C |
MDL Number:
MFCD00008738 CAS Number: 1136-86-3 EINECS: 214-501-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...