p-Tolylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0882-25G T0882-250G
TCT0882-025GEA 74.26 CAD
TCT0882-025G TCT0882-250G
p-Tolylacetic acid ≥98.0% (by GC, titration analysis)
p-Tolylacetic acid

CAS Number: 622-47-9
MDL Number: MFCD00004353
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 267
Melting point (°C): 92


Other Product Names / Synonyms: 4-Methylphenylacetic Acid

Formula: (C₆H₄(CH₃))CH₂COOH
Boiling Pt: 267 °C
Melting Pt: 92 °C
MDL Number: MFCD00004353
CAS Number: 622-47-9
EINECS: 210-738-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7adea78c621ae9b55bd275bd5e72c21ccabcde6f&vid=7adea78c621ae9b55bd275bd5e72c21ccabcde6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR