o-Tolylacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T0881-250G
T0881-25G
TCT0881-250GEA
205.4
CAD
TCT0881-250G
TCT0881-025G
o-Tolylacetic acid ≥98.0% (by titrimetric analysis)
o-Tolylacetic acid
CAS Number: 644-36-0
MDL Number: MFCD00004328
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 89
Other Product Names / Synonyms: 2-Methylphenylacetic Acid
Formula:
(C₆H₄CH₃)CH₂COOH Boiling Pt: 283 °C (1013 hPa) Melting Pt: 89 °C |
MDL Number:
MFCD00004328 CAS Number: 644-36-0 EINECS: 211-416-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...