Thiophene-3-carbaldehyde ≥98.0%

Supplier: TCI America

T0864-5G T0864-25G
TCT0864-005GEA 69.52 CAD
TCT0864-005G TCT0864-025G
Thiophene-3-carbaldehyde ≥98.0%
Thiophene-3-carbaldehyde

CAS Number: 498-62-4
MDL Number: MFCD00005466
Molecular Formula: C5H4OS
Molecular Weight: 112.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 198
Flash Point (°C): 75
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C

Formula: C₅H₄OS
Boiling Pt: 198 °C
Melting Pt: –30 °C
Density: 1.23 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00005466
CAS Number: 498-62-4
EINECS: 207-865-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d5af50a4cede02aed74f1a657ea0f6d3a0d8d47&vid=3d5af50a4cede02aed74f1a657ea0f6d3a0d8d47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR