Tris(4-methylphenyl)phosphine ≥96.0%

Supplier: TCI America

T0862-5G T0862-25G
TCT0862-005GEA 53.72 CAD
TCT0862-005G TCT0862-025G
Tris(4-methylphenyl)phosphine ≥96.0%
Tris(4-methylphenyl)phosphine

CAS Number: 1038-95-5
MDL Number: MFCD00008542
Molecular Formula: C21H21P
Molecular Weight: 304.37
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 149


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Tris(4-methylphenyl)phosphine

Formula: C₂₁H₂₁P
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 149 °C
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00008542
CAS Number: 1038-95-5
EINECS: 213-863-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f7f7cc60dad25dfe34104ade91e6a12a54e71fd&vid=9f7f7cc60dad25dfe34104ade91e6a12a54e71fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR