Tris(4-methoxyphenyl)phosphine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tris(p-methoxyphenyl)phosphine

T0861-25G T0861-5G
TCT0861-25GEA 513.5 CAD
TCT0861-25G TCT0861-005G
Tris(4-methoxyphenyl)phosphine ≥97.0% (by titrimetric analysis)
Tris(4-methoxyphenyl)phosphine

CAS Number: 855-38-9
MDL Number: MFCD00014896
Molecular Formula: C21H21O3P
Molecular Weight: 352.37
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 133


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction), Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Tris(p-anisyl)phosphine

Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Boiling Pt: 446 °C (1013 hPa)
Melting Pt: 133 °C
MDL Number: MFCD00014896
CAS Number: 855-38-9
EINECS: 212-723-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=339b2569511d88f2942002694667e45abd8633a2&vid=339b2569511d88f2942002694667e45abd8633a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR