2,4,6-Triaminopyrimidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T0834-500G T0834-25G
TCT0834-500GEA 785.26 CAD
TCT0834-500G TCT0834-025G
2,4,6-Triaminopyrimidine ≥98.0% (by titrimetric analysis)
2,4,6-Triaminopyrimidine

CAS Number: 1004-38-2
MDL Number: MFCD00006100
Molecular Formula: C4H7N5
Molecular Weight: 125.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 249

Formula: C₄H₇N₅
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 249 °C
Density: 1.45 g/cm³ (20 °C)
MDL Number: MFCD00006100
CAS Number: 1004-38-2
EINECS: 213-720-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27026141779d8a252b42a0c8e96f58e5ec6fa2b0&vid=27026141779d8a252b42a0c8e96f58e5ec6fa2b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR