Tetrafluoro-p-benzoquinone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

T0790-1G T0790-5G
TCT0790-001GEA 75.84 CAD
TCT0790-001G TCT0790-005G
Tetrafluoro-p-benzoquinone ≥98.0% (by HPLC, titration analysis)
Tetrafluoro-p-benzoquinone

CAS Number: 527-21-9
MDL Number: MFCD00001592
Molecular Formula: C6F4O2
Molecular Weight: 180.06
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Fluoranil

Formula: C₆F₄O₂
MW: 180.06 g/mol
Boiling Pt: 133
MDL Number: MFCD00001592
CAS Number: 527-21-9
EINECS: 208-411-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2afd389cb437f9c72827b973bcc583edb2d05bf0&vid=2afd389cb437f9c72827b973bcc583edb2d05bf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR