1-Tetradecyne ≥95.0%

Supplier: TCI America

T0761-25ML T0761-5ML
TCT0761-25MLEA 385.52 CAD
TCT0761-25ML TCT0761-005ML
1-Tetradecyne ≥95.0%
1-Tetradecyne

CAS Number: 765-10-6
MDL Number: MFCD00015076
Molecular Formula: C14H26
Molecular Weight: 194.36
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 252
Melting point (°C): 0
Flash Point (°C): 93
Specific Gravity (20/20): 0.79

Formula: C₁₄H₂₆
Boiling Pt: 252 °C
Density: 0.79 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00015076
CAS Number: 765-10-6
EINECS: 212-139-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1d3c50d169f8faea1b7e326904ac053b7309d8&vid=af1d3c50d169f8faea1b7e326904ac053b7309d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR