TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 98,0

Supplier: TCI America

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

T0740-500G T0740-25G
TCT0740-500GEA 226.62 CAD
TCT0740-500G TCT0740-025G
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 98,0
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)

CAS Number: 1185-53-1
MDL Number: MFCD00012590
Molecular Formula: C4H11NO3
Molecular Weight: 157.59
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White


Product Categories: Bioscience, Buffer-related Reagents
Other Product Names / Synonyms: 2-Amino-2-(hydroxymethyl)-1,3-propanediol Hydrochloride, Trimethylolaminomethane Hydrochloride, Tris Hydrochloride

Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d33b421f63da933d31a6e189ea94aa4a9ac736&vid=99d33b421f63da933d31a6e189ea94aa4a9ac736&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR