1,2,4,5-Tetramethylbenzene ≥98.0%

Supplier: TCI America

T0714-500G T0714-25G
TCT0714-500GEA 570.38 CAD
TCT0714-500G TCT0714-025G
1,2,4,5-Tetramethylbenzene ≥98.0%
1,2,4,5-Tetramethylbenzene

CAS Number: 95-93-2
MDL Number: MFCD00008528
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 197
Melting point (°C): 80
Flash Point (°C): 74


Other Product Names / Synonyms: Durene

Formula: C₁₀H₁₄
Boiling Pt: 197 °C
Melting Pt: 80 °C
Density: 1.036 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00008528
CAS Number: 95-93-2
EINECS: 202-465-7
UN: 1325
ADR: 4.1,II
Merck Index: 13,03501
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4ac9fad5abf10a1513055620e8be1a60dbf6ef1&vid=94aa500e015fe8a88530aaded54bb936df2f007b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR