3,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T0705-25G
T0705-500G
TCT0705-025GEA
36.26
CAD
TCT0705-025G
TCT0705-500G
3,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
3,4,5-Trimethoxybenzoic acid
CAS Number: 118-41-2
MDL Number: MFCD00002501
Molecular Formula: C10H12O5
Molecular Weight: 212.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 227
Melting point (°C): 170
Other Product Names / Synonyms: Gallic Acid Trimethyl Ether
Formula:
C₁₀H₁₂O₅ Boiling Pt: 227 °C Melting Pt: 170 °C Density: 1.31 g/cm³ (24.95 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002501 CAS Number: 118-41-2 EINECS: 204-248-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...