Tetrapropylammonium bromide ≥98.0%

Supplier: TCI America

Synonyms: TPABr

T0694-500G T0694-25G
TCT0694-500GEA 234.79 CAD
TCT0694-500G TCT0694-025G
Tetrapropylammonium bromide ≥98.0%
Tetrapropylammonium bromide

CAS Number: 1941-30-6
MDL Number: MFCD00011840
Molecular Formula: C12H28BrN
Molecular Weight: 266.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₁₂H₂₈BrN
MW: 266.27 g/mol
Melting Pt: 265…274 °C
MDL Number: MFCD00011840
CAS Number: 1941-30-6
EINECS: 217-727-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=432cb826b9a3513ab83a5cfc436b91e3ad02d9c2&vid=432cb826b9a3513ab83a5cfc436b91e3ad02d9c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR