TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 98,0

Supplier: TCI America
Warning

Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic acid, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

T0683-250G T0683-25G
TCT0683-250GEA 1477.3 CAD
TCT0683-250G TCT0683-025G
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 98,0
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)

[Good's buffer component for biological research]
CAS Number: 7365-44-8
MDL Number: MFCD00007532
Molecular Formula: C6H15NO6S
Molecular Weight: 229.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 216


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: TES

Formula: C₆H₁₅NO₆S
MW: 229.25 g/mol
Melting Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00007532
CAS Number: 7365-44-8
EINECS: 230-906-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b95b4cdcb0fb363f55242c0c87b1fb0952281bd2&vid=b95b4cdcb0fb363f55242c0c87b1fb0952281bd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR