Tris(2-hydroxyethyl)-1,3,5-triazinetrione ≥98.0%

Supplier: TCI America

Synonyms: 1,3,5-Tris(2-hydroxyethyl)isocyanuric acid, 1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione, 1,3,5-Tris(2-hydroxyethyl)isocyanurate

T0674-500G T0674-25G
TCT0674-500GEA 169.06 CAD
TCT0674-500G TCT0674-025G
Tris(2-hydroxyethyl)-1,3,5-triazinetrione ≥98.0%
Tris(2-hydroxyethyl)-1,3,5-triazinetrione

CAS Number: 839-90-7
MDL Number: MFCD00003549
Molecular Formula: C9H15N3O6
Molecular Weight: 261.23
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 136
Flash Point (°C): 227


Other Product Names / Synonyms: Isocyanuric Acid Tris(2-hydroxyethyl) Ester

Formula: C₉H₁₅N₃O₆
MW: 261.23 g/mol
Boiling Pt: 527
Melting Pt: 136 °C
Flash Pt: 227 °C
MDL Number: MFCD00003549
CAS Number: 839-90-7
EINECS: 212-660-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1932d689a9df4335be6ada56bdaac62bbbe3b205&vid=1932d689a9df4335be6ada56bdaac62bbbe3b205&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR