Triphenylsilane ≥96.0%
Supplier: TCI America
|
Warning
|
T0661-25G
T0661-5G
TCT0661-025GEA
224.36
CAD
TCT0661-025G
TCT0661-5G
Triphenylsilane ≥96.0%
Triphenylsilane
CAS Number: 789-25-3
MDL Number: MFCD00003003
Molecular Formula: C18H16Si
Molecular Weight: 260.41
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 184
Melting point (°C): 46
Flash Point (°C): 76
Formula:
C₁₈H₁₆Si Boiling Pt: 184 °C Melting Pt: 46 °C Density: 1.152 g/cm³ (20 °C) Flash Pt: 76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003003 CAS Number: 789-25-3 EINECS: 212-333-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...