(±)-2-Tridecanol ≥98.0%

Supplier: TCI America

T0633-25G T0633-5G
TCT0633-025GEA 1943.87 CAD
TCT0633-025G TCT0633-5G
(±)-2-Tridecanol ≥98.0%
(±)-2-Tridecanol

CAS Number: 1653-31-2
MDL Number: MFCD00037601
Molecular Formula: C13H28O
Molecular Weight: 200.37
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 265
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: 2-Tridecyl Alcohol

Formula: C₁₃H₂₈O
Boiling Pt: 265 °C
Melting Pt: 23 °C
Density: 0.83 (20 °C)
MDL Number: MFCD00037601
CAS Number: 1653-31-2
EINECS: 216-723-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa42373e68e1cda3d7a0604d79bee70bce208bdb&vid=aa42373e68e1cda3d7a0604d79bee70bce208bdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR