n-Tetratetracontane ≥97.0%
Supplier: TCI America
T0629-100MG
T0629-1G
TCT0629-100MGEA
55.3
CAD
TCT0629-100MG
TCT0629-001G
n-Tetratetracontane ≥97.0%
n-Tetratetracontane
CAS Number: 7098-22-8
MDL Number: MFCD00015268
Molecular Formula: C44H90
Molecular Weight: 619.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 86
Flash Point (°C): 93
Formula:
H₃C(CH₂)₄₂CH₃ Melting Pt: 86 °C Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015268 CAS Number: 7098-22-8 EINECS: 230-407-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...