Tetrabromo-p-benzoquinone ≥98.0%

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrabromo-1,4-benzoquinone, 2,3,5,6-Tetrabromocyclohexa-2,5-diene-1,4-dione, p-Bromanil, Tetrabromoquinone, 4-Bromanil, Tetrabromo-1,4-benzoquinone

T0617-25G T0617-5G
TCT0617-025GEA 417.12 CAD
TCT0617-025G TCT0617-005G
Tetrabromo-p-benzoquinone ≥98.0%
Tetrabromo-p-benzoquinone

CAS Number: 488-48-2
MDL Number: MFCD00013785
Molecular Formula: C6Br4O2
Molecular Weight: 423.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow


Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Tetrabromo-1,4-benzoquinone

Formula: C₆Br₄O₂
MW: 423.68 g/mol
MDL Number: MFCD00013785
CAS Number: 488-48-2
EINECS: 207-679-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59616a476a74221bad67e76f40f02cba59bf8aeb&vid=59616a476a74221bad67e76f40f02cba59bf8aeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR