Bis(cyclopentadienyl)titanium dichloride ≥98.0%

Supplier: TCI America

Synonyms: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

T0616-5G T0616-25G
TCT0616-5GEA 42.66 CAD
TCT0616-5G TCT0616-025G
Bis(cyclopentadienyl)titanium dichloride ≥98.0%
Bis(cyclopentadienyl)titanium dichloride

CAS Number: 1271-19-8
MDL Number: MFCD00003723
Molecular Formula: C10H10Cl2Ti
Molecular Weight: 248.96
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow Red


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Bis(cyclopentadienyl)titanium Dichloride

Formula: C₁₀H₁₀Cl₂Ti
MW: 248.97 g/mol
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41fe014061709555d928ec48876f10c1e4b13bba&vid=41fe014061709555d928ec48876f10c1e4b13bba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR