Farnesol (mixture of isomers)

Supplier: TCI America

T0608-5G T0608-25G T0608-100G
TCT0608-005GEA 35.38 CAD
TCT0608-005G TCT0608-025G TCT0608-100G
Farnesol (mixture of isomers)
Farnesol (mixture of isomers)

CAS Number: 4602-84-0
MDL Number: MFCD00002918
Molecular Formula: C15H26O
Molecular Weight: 222.37
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 143
Flash Point (°C): 96
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: 3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol

Formula: C₁₅H₂₆O
Boiling Pt: 143 °C
Density: 0.89 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00002918
CAS Number: 4602-84-0
EINECS: 225-004-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60a4bcb2abeedc288b290f286eba6e40c6e4acf7&vid=60a4bcb2abeedc288b290f286eba6e40c6e4acf7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR