α-Tropolone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Tropolone, 2-Hydroxytropone, 2-Hydroxycyclohepta-2,4,6-trien-1-one, 1,2-Tropolone, 2-Hydroxycyclohepta-2,4,6-trienone, α-Tropolone

T0606-1G T0606-5G T0606-25G
TCT0606-001GEA 42.66 CAD
TCT0606-001G TCT0606-005G TCT0606-25G
α-Tropolone ≥98.0% (by GC, titration analysis)
α-Tropolone

CAS Number: 533-75-5
MDL Number: MFCD00004158
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 53
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Hydroxy-2,4,6-cycloheptatrien-1-one

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 66…67 °C (0.1 mmHg)
Melting Pt: 53 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS Number: 533-75-5
EINECS: 208-577-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5987d60925df81e32cc0cff2a3f15579a472046b&vid=5987d60925df81e32cc0cff2a3f15579a472046b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR