Trifluoroacetamide ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,2,2-Trifluoroacetamide

T0598-500G T0598-25G T0598-100G
TCT0598-500GEA 284.49 CAD
TCT0598-500G TCT0598-25G TCT0598-100G
Trifluoroacetamide ≥98.0%
Trifluoroacetamide

CAS Number: 354-38-1
MDL Number: MFCD00008008
Molecular Formula: C2H2F3NO
Molecular Weight: 113.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 163
Melting point (°C): 72

Formula: C₂H₂F₃NO
MW: 113.04 g/mol
Boiling Pt: 163 °C
Melting Pt: 72 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00008008
CAS Number: 354-38-1
EINECS: 206-559-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc1839f20dac18b22e4e542e2c21687f8caba61b&vid=dc1839f20dac18b22e4e542e2c21687f8caba61b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR