n-Triacontane ≥98.0%

Supplier: TCI America

T0594-5G T0594-25G
TCT0594-005GEA 317.72 CAD
TCT0594-005G TCT0594-025G
n-Triacontane ≥98.0%
n-Triacontane

CAS Number: 638-68-6
MDL Number: MFCD00009410
Molecular Formula: C30H62
Molecular Weight: 422.83
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 200
Melting point (°C): 66
Flash Point (°C): 166

Formula: H₃C(CH₂)₂₈CH₃
Boiling Pt: 200 °C
Melting Pt: 66 °C
Density: 0.7797 g/cm³ (20 °C)
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00009410
CAS Number: 638-68-6
EINECS: 211-349-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28d6b1381ad7fc8f788c31b9d9b4affe76677673&vid=08a845dd4190bb5666172079188310a90d015140&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR