2-Tolualdehyde ≥98.0%

Supplier: TCI America

T0566-25G T0566-500G T0566-100G
TCT0566-025GEA 74.29 CAD
TCT0566-025G TCT0566-500G TCT0566-100G
2-Tolualdehyde ≥98.0%
2-Tolualdehyde

CAS Number: 529-20-4
MDL Number: MFCD00003338
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 200
Flash Point (°C): 67
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-Methylbenzaldehyde

Formula: C₈H₈O
Boiling Pt: 200 °C
Melting Pt: –35 °C
Density: 1.04 (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Merck Index: 12,09665
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce8e4a9e7605c01ea16f1cf58a9735b1a02d36be&vid=d926b7449fda11f0ace6349d6cccc6440f8b1fdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR