Rubrene ≥98.0%

Supplier: TCI America

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

T0561-1G T0561-100MG
TCT0561-1GEA 153.26 CAD
TCT0561-1G TCT0561-100MG
Rubrene ≥98.0%
Rubrene

CAS Number: 517-51-1
MDL Number: MFCD00003703
Molecular Formula: C42H28
Molecular Weight: 532.69
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Yellow
Melting point (°C): 331


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Rubrene

Formula: C₄₂H₂₈
MW: 532.68 g/mol
Melting Pt: 331 °C
MDL Number: MFCD00003703
CAS Number: 517-51-1
EINECS: 208-242-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93e824ee9511d447cca2b0b432eccf8e11646ad9&vid=93e824ee9511d447cca2b0b432eccf8e11646ad9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR