L(-)-Tyrosine ≥98.5% (by titrimetric analysis)
Supplier: TCI America
T0550-500G
T0550-25G
T0550-100G
TCT0550-500GEA
886.38
CAD
TCT0550-500G
TCT0550-025G
TCT0550-100G
L(-)-Tyrosine ≥98.5% (by titrimetric analysis)
L(-)-Tyrosine
CAS Number: 60-18-4
MDL Number: MFCD00002606
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -12 deg (C=5, 1mol/L HCl)
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Tyr-OH
Formula:
C₉H₁₁NO₃ Boiling Pt: 351 °C (1013 hPa) Melting Pt: 285 °C Density: 1.41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002606 CAS Number: 60-18-4 EINECS: 200-460-4 Merck Index: 13,09907 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...