Tween® 80 (Polysorbate)

Supplier: TCI America

T0546-25G T0546-500G
TCT0546-025GEA 67.59 CAD
TCT0546-025G TCT0546-500G
Tween® 80 (Polysorbate)
Tween® 80 (Polysorbate)

CAS Number: 9005-65-6
MDL Number: MFCD00082107
Form: Clear Liquid
Color: Slightly Pale Yellow
Melting point (°C): -25
Flash Point (°C): 295
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monooleate

Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.08 (20 °C)
Flash Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9
Merck Index: 13,07664

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR