Tween® 60 (Polysorbate)
Supplier: TCI America
T0545-25G
T0545-500G
TCT0545-025GEA
33.18
CAD
TCT0545-025G
TCT0545-500G
Tween® 60 (Polysorbate)
Tween® 60 (Polysorbate)
CAS Number: 9005-67-8
MDL Number: MFCD01780110
Form: Crystal
Color: Slightly Pale Yellow
Flash Point (°C): 277
Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monostearate
Formula:
C₆₄H₁₂₆O₂₆ Boiling Pt: ∼110 °C (1013 hPa) Density: 1.08 g/cm³ (20 °C) Flash Pt: 277 °C Storage Temperature: Ambient |
MDL Number:
MFCD01780110 CAS Number: 9005-67-8 EINECS: 500-020-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...