Tween® 40 (Polysorbate)

Supplier: TCI America

T0544-25G T0544-500G
TCT0544-025GEA 33.59 CAD
TCT0544-025G TCT0544-500G
Tween® 40 (Polysorbate)
Tween® 40 (Polysorbate)

CAS Number: 9005-66-7
MDL Number: MFCD00165345
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 268


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monopalmitate

Formula: C₆₂H₁₂₂O₂₆
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: ∼0.1 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 268 °C
Storage Temperature: Ambient
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR