Tween® 20 (Polysorbate)

Supplier: TCI America

T0543-25G T0543-500G
TCT0543-025GEA 40.35 CAD
TCT0543-025G TCT0543-500G
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)

CAS Number: 9005-64-5
MDL Number: MFCD00165986
Form: Clear Liquid
Color: Yellow
Melting point (°C): -14
Flash Point (°C): 297
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monolaurate

Formula: C₅₈H₁₁₄O₂₆
Melting Pt: –14 °C
Density: 1.11 (20 °C)
Flash Pt: 297 °C
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR