D-(+)-Tryptophan ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T0539-25G T0539-5G
TCT0539-025GEA 178.54 CAD
TCT0539-025G TCT0539-005G
D-(+)-Tryptophan ≥98.0% (by HPLC, titration analysis)
D(+)-Tryptophan

CAS Number: 153-94-6
MDL Number: MFCD00005647
Molecular Formula: C11H12N2O2
Molecular Weight: 204.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 31 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-(+)-2-Amino-3-(3-indolyl)propionic Acid, H-D-Trp-OH

Formula: C₁₁H₁₂N₂O₂
Melting Pt: 280…283 °C
MDL Number: MFCD00064339
CAS Number: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b47c38ace9ddeb2a66a1015a2a110d7b543a026&vid=6b47c38ace9ddeb2a66a1015a2a110d7b543a026&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR