2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: 2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine

T0530-1G T0530-25G T0530-5G
TCT0530-001GEA 51.19 CAD
TCT0530-001G TCT0530-025G 76220-486
2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98.0%
2,4,6-Tris(2-pyridyl)-1,3,5-triazine

CAS Number: 3682-35-7
MDL Number: MFCD00006045
Molecular Formula: C18H12N6
Molecular Weight: 312.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Ligands, Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: TPTZ, 2,4,6-Tris(2-pyridyl)-1,3,5-triazine

Formula: C₁₈H₁₂N₆
MW: 312.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006045
CAS Number: 3682-35-7
EINECS: 222-965-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff67a6dad76d56a73e075574ae143a6aa863da4&vid=8ff67a6dad76d56a73e075574ae143a6aa863da4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR