Triphenylphosphine ≥95.0%

Supplier: TCI America
Warning

T0519-25G T0519-500G T0519-100G
TCT0519-025GEA 37.89 CAD
TCT0519-025G TCT0519-500G TCT0519-100G
Triphenylphosphine ≥95.0%
Triphenylphosphine

CAS Number: 603-35-0
MDL Number: MFCD00003043
Molecular Formula: C18H15P
Molecular Weight: 262.29
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 260
Melting point (°C): 81
Flash Point (°C): 187

Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377
Melting Pt: 79
Density: 1.194 g/cm³ (20 °C)
Flash Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebe0c3e17ddb3c6f12093e1915d0d56d67894eb&vid=aebe0c3e17ddb3c6f12093e1915d0d56d67894eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR