Triphenylene ≥96.0%

Supplier: TCI America
Danger

T0513-1G T0513-100MG
TCT0513-001GEA 51.46 CAD
TCT0513-001G TCT0513-100MG
Triphenylene ≥96.0%
Triphenylene

CAS Number: 217-59-4
MDL Number: MFCD00001108
Molecular Formula: C18H12
Molecular Weight: 228.29
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 425
Melting point (°C): 199
Lambda max.: 335 nm (Hexane)


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: 9,10-Benzophenanthrene, 1,2:3,4-Dibenznaphthalene, Isochrysene

Formula: C₁₈H₁₂
Boiling Pt: 425 °C
Melting Pt: 199 °C
Density: 1.302 g/cm³ (20 °C)
MDL Number: MFCD00001108
CAS Number: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9,III
Merck Index: 12,09870
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70150a32e28ffeea4b05475857393c4e5a78b30b&vid=70150a32e28ffeea4b05475857393c4e5a78b30b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR