Triphenylamine ≥98.0%

Supplier: TCI America

T0507-250G T0507-25G
TCT0507-250GEA 486.64 CAD
TCT0507-250G TCT0507-025G
Triphenylamine ≥98.0%
Triphenylamine

CAS Number: 603-34-9
MDL Number: MFCD00003020
Molecular Formula: C18H15N
Molecular Weight: 245.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 127

Formula: C₁₈H₁₅N
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 127 °C
Density: 0.774 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003020
CAS Number: 603-34-9
EINECS: 210-035-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=598aee6414624881a8a0433b2445fdc074f6cbff&vid=8f7e54d3e61abd8d775dd33506fff8539ee88910&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR