Trioctylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Tri-n-octylamine
T0502-25ML
T0502-500ML
T0502-100ML
TCT0502-025MLEA
39.5
CAD
TCT0502-025ML
TCT0502-500ML
TCT0502-100ML
Trioctylamine ≥97.0% (by GC, titration analysis)
Trioctylamine
CAS Number: 1116-76-3
MDL Number: MFCD00009560
Molecular Formula: C24H51N
Molecular Weight: 353.68
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 367
Flash Point (°C): 184
Specific Gravity (20/20): 0.81
Formula:
C₂₄H₅₁N MW: 353.68 g/mol Boiling Pt: 367 °C Melting Pt: –39 °C Density: 0.81 (20 °C) Flash Pt: 184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009560 CAS Number: 1116-76-3 EINECS: 214-242-1 UN: 3082 ADR: 9,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...