(±)-2,2,4-Trimethyl-1,3-pentanediol ≥97.0%

Supplier: TCI America

T0483-25G T0483-500G
TCT0483-025GEA 47.4 CAD
TCT0483-025G TCT0483-500G
(±)-2,2,4-Trimethyl-1,3-pentanediol ≥97.0%
(±)-2,2,4-Trimethyl-1,3-pentanediol

CAS Number: 144-19-4
MDL Number: MFCD00004681
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 232
Melting point (°C): 54
Flash Point (°C): 140

Formula: C₈H₁₈O₂
Boiling Pt: 232 °C
Melting Pt: 54 °C
Density: 0.9485 g/cm³ (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00004681
CAS Number: 144-19-4
EINECS: 205-619-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35dd4cfc50ef0d8e26a2ccdce2353da3dc13b729&vid=35dd4cfc50ef0d8e26a2ccdce2353da3dc13b729&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR