2,2,4-Trimethylpentane ≥99.0%

Supplier: TCI America

Synonyms: IsobutyltrimethylmethaneIsooctaneIsobutyltrimethylethane

T0481-500ML T0481-25ML
TCT0481-25MLEA 31.22 CAD
TCT0481-25ML TCT0481-500ML
2,2,4-Trimethylpentane ≥99.0%
2,2,4-Trimethylpentane

CAS Number: 540-84-1
MDL Number: MFCD00008943
Molecular Formula: C8H18
Molecular Weight: 114.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 99
Flash Point (°C): -9
Specific Gravity (20/20): 0.69


Other Product Names / Synonyms: Isooctane

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99 °C
Melting Pt: –107 °C
Density: 0.69 (20 °C)
Flash Pt: –9 °C
MDL Number: MFCD00008943
CAS Number: 540-84-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://ca.vwr.com/stibo/web/std.lang.all/47/34/28994734.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR