1,1,1-Tris(hydroxymethyl)propane ≥98.0%

Supplier: TCI America

T0480-25G T0480-500G
TCT0480-25GEA 32.45 CAD
TCT0480-25G TCT0480-500G
1,1,1-Tris(hydroxymethyl)propane ≥98.0%
1,1,1-Tris(hydroxymethyl)propane

CAS Number: 77-99-6
MDL Number: MFCD00004694
Molecular Formula: C6H14O3
Molecular Weight: 134.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 149
Melting point (°C): 58
Flash Point (°C): 172


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,2-Bis(hydroxymethyl)-1-butanol, 2-Ethyl-2-hydroxymethyl-1,3-propanediol, Hexaglycerol, 1,1,1-Tris(hydroxymethyl)propane

Formula: C₆H₁₄O₃
Boiling Pt: 149 °C
Melting Pt: 58 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3413a080cbfe27f89b3efaf9622596c6b4250232&vid=3413a080cbfe27f89b3efaf9622596c6b4250232&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR