1,1,1-Tris(hydroxymethyl)ethane ≥98.0%
Supplier: TCI America
Synonyms:
2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol
CAS Number: 77-85-0
MDL Number: MFCD00004687
Molecular Formula: C5H12O3
Molecular Weight: 120.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 137
Melting point (°C): 201
Flash Point (°C): 160
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-Hydroxymethyl-2-methyl-1,3-propanediol, Pentaglycerol, 1,1,1-Tris(hydroxymethyl)ethane
Formula:
C₅H₁₂O₃ MW: 120.15 g/mol Boiling Pt: 137 °C Melting Pt: 201 °C Flash Pt: 160 °C |
MDL Number:
MFCD00004687 CAS Number: 77-85-0 EINECS: 201-063-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...