Trimethylammonium chloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
|
Warning
|
T0465-25G
T0465-500G
T0465-100G
TCT0465-025GEA
36.34
CAD
TCT0465-025G
TCT0465-500G
TCT0465-100G
Trimethylammonium chloride ≥98.0% (by total nitrogen basis)
Trimethylammonium chloride
CAS Number: 593-81-7
MDL Number: MFCD00012478
Molecular Formula: C3H9N
Molecular Weight: 95.57
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Formula:
C₃H₁₀ClN Melting Pt: 273…278 °C Density: ∼1 g/cm³ (20 °C) |
MDL Number:
MFCD00012478 CAS Number: 593-81-7 EINECS: 209-810-0 Merck Index: 12,09841 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...